Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014062.2 | NP_054781.1 | 9 | 1560 | ||
NR_074074.1 | 8 | 1429 | ||||
GenBank mRNA | AB026125.1 | - | 9 | 1518 | ||
AF190161.1 | - | 9 | 1487 | |||
AK001028.1 | - | 9 | 1515 | |||
AK126196.1 | - | 1 | 3045 | |||
AY487344.1 | - | 9 | 1534 | |||
BC000050.2 | - | 6 | 1147 | |||
BC064630.1 | - | 9 | 1503 | 5740890 | ||
BM793847.1 | - | 1 | 244 |