Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014329.4 | NP_055144.3 | 3 | 497 | ||
GenBank mRNA | AK128582.1 | - | 4 | 568 | ||
AK291049.1 | - | 3 | 265 | |||
AK302563.1 | - | 3 | 263 | |||
BC043616.1 | - | 3 | 400 | 6065418 | ||
BC064567.1 | - | 3 | 394 | 5575195 | ||
BX445706.2 | - | 3 | 449 | |||
L26339.1 | - | 3 | 394 |