Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBGCP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014444.4 | NP_055259.2 | 12, 13 | 1567 | ||
XM_017022078.1 | XP_016877567.1 | 12, 13 | 1567 | |||
GenBank mRNA | AK127306.1 | - | 2, 3 | 1711 | ||
BC009870.2 | - | 12, 13 | 1492 | 3940649 | ||
BC012801.2 | - | 12, 13 | 1511 | 2821891 |