Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023567.4 | NP_001018861.3 | 16 | 3360 | ||
NR_027410.1 | 24 | 5608 | ||||
XM_011521596.1 | XP_011519898.1 | 23 | 4205 | |||
XM_011521597.1 | XP_011519899.1 | 22 | 4156 | |||
XM_017022210.1 | XP_016877699.1 | 24 | 4509 | |||
XM_017022211.1 | XP_016877700.1 | 24 | 4278 | |||
XM_017022212.1 | XP_016877701.1 | 23 | 4832 | |||
XM_017022213.1 | XP_016877702.1 | 23 | 4234 | |||
XM_017022214.1 | XP_016877703.1 | 24 | 4506 | |||
XM_017022215.1 | XP_016877704.1 | 24 | 5031 | |||
XM_017022216.1 | XP_016877705.1 | 23 | 5629 | |||
XM_017022217.1 | XP_016877706.1 | 22 | 5551 | |||
XM_017022218.1 | XP_016877707.1 | 24 | 5658 | |||
GenBank mRNA | AL133593.1 | - | 1 | 331 | ||
BX647673.1 | - | 23 | 5576 |
Gene Symbol: GOLGA8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181077.3 | NP_851422.1 | 16 | 3358 | ||
NR_027409.1 | 24 | 5620 | ||||
XM_006720440.2 | XP_006720503.1 | 16 | 3323 | |||
XM_017022018.1 | XP_016877507.1 | 20 | 5754 | |||
XM_017022019.1 | XP_016877508.1 | 19 | 5450 | |||
XM_017022020.1 | XP_016877509.1 | 14 | 3445 | |||
GenBank mRNA | AB020662.1 | - | 15 | 3338 | ||
AF204231.1 | - | 15 | 3675 | |||
AL117569.1 | - | 2 | 1665 | |||
AL137686.1 | - | 15 | 3079 | |||
BC064341.1 | - | 1 | 790 | |||
BC150329.1 | - | 15 | 3349 | |||
BC152410.1 | - | 15 | 3338 | |||
BX648160.1 | - | 23 | 5574 |