Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286420.1 | NP_001273349.1 | 4 | 593 | ||
NM_016454.3 | NP_057538.1 | 5 | 754 | |||
GenBank mRNA | AF151018.1 | - | 5 | 707 | ||
AK075227.1 | - | 5 | 733 | |||
AK290524.1 | - | 4 | 577 | |||
BC002583.2 | - | 4 | 558 | 3161718 | ||
DB506932.1 | - | 2 | 165 |