Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BANP SIRNA Assays ›
Gene Symbol:
BANP
RRN3P2
Gene Name:
BTG3 associated nuclear protein
RRN3 homolog, RNA polymerase I transcription factor pseudogene 2
Gene Aliases:
BEND1, SMAR1, SMARBP1
-
Chromosome Location:
Chr.16: 87949216 - 88077318 on Build GRCh38
Chromosome Location:
Chr.16: 29074842 - 29116718 on Build GRCh38
UniGene ID:
Hs.461705
Hs.720348
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
140455
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BANP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001173539.1
NP_001167010.1
3
265
NM_001173540.1
NP_001167011.1
3
283
NM_001173541.1
NP_001167012.1
3
265
NM_001173542.1
NP_001167013.1
3
304
NM_001173543.1
NP_001167014.1
3
334
NM_017869.3
NP_060339.2
3
265
NM_079837.2
NP_524576.2
3
265
XM_005256008.3
XP_005256065.1
3
281
XM_005256009.3
XP_005256066.1
3
263
XM_011523170.2
XP_011521472.1
3
281
XM_011523172.2
XP_011521474.1
4
526
XM_011523174.2
XP_011521476.1
3
1052
XM_011523175.2
XP_011521477.1
3
683
XM_011523176.2
XP_011521478.1
3
745
XM_011523177.2
XP_011521479.1
3
1253
XM_011523178.2
XP_011521480.1
3
281
XM_011523179.2
XP_011521481.1
3
263
XM_011523180.2
XP_011521482.1
4
345
XM_011523181.2
XP_011521483.1
3
281
XM_011523182.1
XP_011521484.1
3
281
XM_011523183.2
XP_011521485.1
3
258
XM_011523184.2
XP_011521486.1
3
214
XM_011523186.2
XP_011521488.1
1
43
XM_017023378.1
XP_016878867.1
4
385
XM_017023379.1
XP_016878868.1
3
665
XM_017023380.1
XP_016878869.1
3
265
XM_017023381.1
XP_016878870.1
3
283
XM_017023383.1
XP_016878872.1
3
230
GenBank mRNA
AK001039.1
-
3
265
AK094158.1
-
3
335
AK125232.1
-
3
263
AK293472.1
-
3
227
AK297945.1
-
3
237
AK315089.1
-
3
263
AK315535.1
-
3
304
BC009424.1
-
3
210
3530821
DA632793.1
-
3
265
KU178664.1
-
2
113
KU178665.1
-
2
113
KU178666.1
-
2
113
KU178667.1
-
2
113
KU178668.1
-
2
113
KU178669.1
-
2
113
KU178670.1
-
2
135
Gene Symbol: RRN3P2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
AK225670.1
-
16
1606
AK298442.1
-
14
1497
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
cell cycle
multicellular organism development
covalent chromatin modification
protein localization to nucleus
negative regulation of protein catabolic process
positive regulation of transcription, DNA-templated
regulation of signal transduction by p53 class mediator
p53 binding
DNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0