Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161575.1 | NP_001155047.1 | 5 | 846 | ||
NM_018296.5 | NP_060766.5 | 8 | 1116 | |||
XM_005256025.2 | XP_005256082.1 | 8 | 1116 | |||
XM_005256027.2 | XP_005256084.1 | 8 | 1116 | |||
XM_005256028.1 | XP_005256085.1 | 7 | 988 | |||
XM_011523200.1 | XP_011521502.1 | 8 | 1116 | |||
XM_011523201.1 | XP_011521503.1 | 5 | 736 | |||
XM_011523202.2 | XP_011521504.1 | 4 | 591 | |||
XM_011523204.1 | XP_011521506.1 | 6 | 915 | |||
XM_011523205.1 | XP_011521507.1 | 7 | 1006 | |||
XM_011523206.2 | XP_011521508.1 | 8 | 1116 | |||
XM_017023400.1 | XP_016878889.1 | 8 | 1116 | |||
GenBank mRNA | AK001866.1 | - | 5 | 895 | ||
AK292311.1 | - | 5 | 825 | |||
AK301766.1 | - | 5 | 800 | |||
AY313780.1 | - | 5 | 809 | |||
AY963267.1 | - | 8 | 1116 | |||
BC026156.2 | - | 8 | 1116 | 4828487 |