Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114118.2 | NP_001107590.1 | 12, 13 | 1663 | ||
XM_005256717.2 | XP_005256774.1 | 12, 13 | 1663 | |||
XM_006721556.2 | XP_006721619.1 | 11, 12 | 1552 | |||
XM_011523954.2 | XP_011522256.1 | 12, 13 | 1580 | |||
XM_011523955.2 | XP_011522257.1 | 11, 12 | 1434 | |||
XM_017024826.1 | XP_016880315.1 | 11, 12 | 1550 | |||
XM_017024827.1 | XP_016880316.1 | 6, 7 | 1137 | |||
XM_017024828.1 | XP_016880317.1 | 7, 8 | 955 | |||
XM_017024829.1 | XP_016880318.1 | 7, 8 | 955 | |||
GenBank mRNA | AJ277841.1 | - | 7, 8 | 960 | ||
AK125048.1 | - | 9, 10 | 2352 | |||
AK223470.1 | - | 5, 6 | 839 | |||
BC010707.1 | - | 5, 6 | 778 |