Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144955.1 | NP_001138427.1 | 5 | 914 | ||
NM_020234.5 | NP_064619.2 | 6 | 1042 | |||
XM_017022419.1 | XP_016877908.1 | 5 | 961 | |||
XM_017022420.1 | XP_016877909.1 | 6 | 1823 | |||
XM_017022421.1 | XP_016877910.1 | 6 | 1054 | |||
XM_017022422.1 | XP_016877911.1 | 5 | 1695 | |||
XM_017022427.1 | XP_016877916.1 | 6 | 979 | |||
XM_017022428.1 | XP_016877917.1 | 6 | 984 | |||
XM_017022429.1 | XP_016877918.1 | 6 | 1930 | |||
XM_017022430.1 | XP_016877919.1 | 6 | 1133 | |||
XM_017022431.1 | XP_016877920.1 | 5 | 575 | |||
GenBank mRNA | AF168717.1 | - | 5 | 902 | ||
AK290184.1 | - | 5 | 883 | |||
BC032535.2 | - | 6 | 982 | 5527713 | ||
BC093073.1 | - | 6 | 972 | 30326731 |