Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025099.5 | NP_079375.3 | 17 | 2902 | ||
NR_046431.1 | 17 | 2797 | ||||
XM_006721577.3 | XP_006721640.1 | 16 | 2757 | |||
XM_006721578.3 | XP_006721641.1 | 17 | 2886 | |||
XM_011524010.2 | XP_011522312.1 | 17 | 2781 | |||
XM_011524011.2 | XP_011522313.1 | 16 | 2651 | |||
GenBank mRNA | AK025823.1 | - | 7 | 1045 | ||
AK091077.1 | - | 17 | 2853 | |||
AK225375.1 | - | 7 | 1045 | |||
AK310636.1 | - | 6 | 1071 | |||
AL831955.1 | - | 17 | 2744 | |||
BC026057.2 | - | 8 | 1043 | |||
BC110373.1 | - | 9 | 1269 | |||
BC111783.1 | - | 17 | 2853 | 40054070 | ||
BX406692.2 | - | 4 | 461 | |||
DQ451688.1 | - | 17 | 2843 | |||
FJ790670.1 | - | 17 | 2843 |