Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160227.1 | NP_001153699.1 | 2 | 309 | ||
NM_025137.3 | NP_079413.3 | 2 | 309 | |||
XM_006720700.1 | XP_006720763.1 | 2 | 309 | |||
XM_006720701.3 | XP_006720764.1 | 2 | 309 | |||
XM_017022634.1 | XP_016878123.1 | 2 | 309 | |||
XM_017022635.1 | XP_016878124.1 | 2 | 309 | |||
GenBank mRNA | AB058743.2 | - | 2 | 279 | ||
AB470308.1 | - | 2 | 278 | |||
AB470309.1 | - | 2 | 278 | |||
AB470310.1 | - | 2 | 278 | |||
AL834168.1 | - | 1 | 31 | |||
BC047416.1 | - | 2 | 289 | |||
BC150640.1 | - | 2 | 306 | 9057010 | ||
BC153879.1 | - | 2 | 279 | 8860108 | ||
DA926289.1 | - | 2 | 309 |
Gene Symbol: ISLR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK074668.1 | - | 293 |