Search
Search

FOPNL| Gene Symbol: FOPNL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304499.1 | NP_001291428.1 | 5, 6 | 585 | ||
| NM_001304500.1 | NP_001291429.1 | 3, 4 | 315 | |||
| NM_144600.3 | NP_653201.1 | 4, 5 | 513 | |||
| NR_130755.1 | 3, 4 | 428 | ||||
| NR_130756.1 | 5, 6 | 613 | ||||
| NR_130757.1 | 2, 3 | 230 | ||||
| GenBank mRNA | AK055715.1 | - | 4, 5 | 456 | ||
| AL832498.1 | - | 4, 5 | 476 | |||
| AY507846.1 | - | 4, 5 | 469 | |||
| BC022321.1 | - | 4, 5 | 464 | 4702249 | ||
| BI460301.1 | - | 5, 6 | 551 | |||
| BI550657.1 | - | 3, 4 | 404 | |||
| BU661163.1 | - | 2, 3 | 181 | |||
| CB991926.1 | - | 5, 6 | 602 | |||
| DB103294.1 | - | 3, 4 | 259 | |||