Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145294.4 | NP_660337.3 | 41 | 5442 | ||
XM_017023023.1 | XP_016878512.1 | 42 | 5712 | |||
XM_017023024.1 | XP_016878513.1 | 24 | 3325 | |||
GenBank mRNA | AK091062.1 | - | 14 | 2632 | ||
AK092719.1 | - | 12 | 2158 | |||
AK093609.1 | - | 6 | 1842 | |||
AK131510.1 | - | 24 | 3748 | |||
BC065836.1 | - | 6 | 1354 |