Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DMD SIRNA Assays ›
Gene Symbol:
DMD
Gene Name:
dystrophin
Gene Aliases:
BMD, CMD3B, DXS142, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, MRX85
Chromosome Location:
Chr.X: 31119219 - 33339609 on Build GRCh38
UniGene ID:
Hs.495912
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
14146
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DMD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000109.3
NP_000100.2
66
9906
NM_004006.2
NP_003997.1
66
9830
NM_004009.3
NP_004000.1
66
9837
NM_004010.3
NP_004001.1
66
9920
NM_004011.3
NP_004002.2
38
5608
NM_004012.3
NP_004003.1
38
5751
NM_004013.2
NP_004004.1
23
3247
NM_004014.2
NP_004005.1
12
1460
NM_004015.2
NP_004006.1
5
460
NM_004016.2
NP_004007.1
5
460
NM_004017.2
NP_004008.1
5
460
NM_004018.2
NP_004009.1
5
460
NM_004019.2
NP_004010.1
5
460
NM_004020.3
NP_004011.2
23
3247
NM_004021.2
NP_004012.1
23
3247
NM_004022.2
NP_004013.1
23
3247
NM_004023.2
NP_004014.1
23
3247
XM_006724468.2
XP_006724531.1
66
9783
XM_006724469.3
XP_006724532.1
66
9757
XM_006724470.3
XP_006724533.1
66
9783
XM_006724473.2
XP_006724536.1
65
9645
XM_006724474.3
XP_006724537.1
66
9783
XM_006724475.2
XP_006724538.1
66
9783
XM_011545467.1
XP_011543769.1
65
9660
XM_011545468.2
XP_011543770.1
66
9783
XM_017029328.1
XP_016884817.1
66
9783
XM_017029331.1
XP_016884820.1
26
4359
GenBank mRNA
AB208836.1
-
20
2694
AK299936.1
-
5
502
BC028720.1
-
5
511
BC094758.1
-
5
474
BC127103.1
-
20
2771
BC150141.1
-
20
2771
DB233002.1
-
5
460
KU178030.1
-
5
382
KU178031.1
-
5
382
KU178032.1
-
5
382
KU178033.1
-
5
382
M18533.1
-
66
9794
M92650.1
-
5
434
X14298.1
-
66
9684
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001005246, XM_008773253, XM_008773252, X69767.
This siRNA also targets the following transcripts in Mouse: NM_007868, KJ480729, KJ480728, M68859, AK075809, and others.
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
regulation of heart rate
cytoskeleton organization
muscle organ development
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
regulation of skeletal muscle contraction by regulation of release of sequestered calcium ion
regulation of skeletal muscle contraction
muscle attachment
muscle filament sliding
negative regulation of peptidyl-serine phosphorylation
cellular protein localization
response to muscle stretch
peptide biosynthetic process
cellular protein complex assembly
muscle cell cellular homeostasis
muscle fiber development
cardiac muscle contraction
regulation of ryanodine-sensitive calcium-release channel activity
cardiac muscle cell action potential
regulation of voltage-gated calcium channel activity
negative regulation of peptidyl-cysteine S-nitrosylation
positive regulation of sodium ion transmembrane transporter activity
dystroglycan binding
actin binding
structural constituent of cytoskeleton
protein binding
zinc ion binding
structural constituent of muscle
myosin binding
vinculin binding
nitric-oxide synthase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-p5628:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0