Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANKS1B SIRNA Assays ›
          Gene Symbol:
          ANKS1B
          Gene Name:
          ankyrin repeat and sterile alpha motif domain containing 1B
          Gene Aliases:
          AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2
          Chromosome Location:
          Chr.12: 98729904 - 99984727 on Build GRCh38
          UniGene ID:
          Hs.506458
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 141558

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ANKS1B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001204066.1 NP_001190995.1 2 111
          NM_001204067.1 NP_001190996.1 2 172
          NM_001204068.1 NP_001190997.1 1 455
          NM_001204069.1 NP_001190998.1 1 455
          NM_001204070.1 NP_001190999.1 1 455
          NM_020140.3 NP_064525.1 1 455
          NM_152788.4 NP_690001.3 15 2887
          NM_181670.3 NP_858056.2 1 455
          XM_005269028.4 XP_005269085.1 15 2498
          XM_005269029.4 XP_005269086.1 15 2458
          XM_006719504.3 XP_006719567.1 15 2960
          XM_006719505.3 XP_006719568.1 15 2960
          XM_006719506.3 XP_006719569.1 15 2960
          XM_006719507.3 XP_006719570.1 15 2960
          XM_006719508.3 XP_006719571.1 15 2960
          XM_006719509.3 XP_006719572.1 15 2960
          XM_006719510.3 XP_006719573.1 15 2960
          XM_006719512.3 XP_006719575.1 15 2960
          XM_006719513.3 XP_006719576.1 15 2960
          XM_006719514.3 XP_006719577.1 15 2948
          XM_011538571.2 XP_011536873.1 15 2960
          XM_017019651.1 XP_016875140.1 15 2960
          XM_017019652.1 XP_016875141.1 15 2948
          XM_017019653.1 XP_016875142.1 15 2960
          XM_017019654.1 XP_016875143.1 15 2948
          XM_017019655.1 XP_016875144.1 15 2960
          XM_017019656.1 XP_016875145.1 15 2960
          XM_017019657.1 XP_016875146.1 15 2960
          XM_017019658.1 XP_016875147.1 15 2960
          XM_017019659.1 XP_016875148.1 1 451
          XM_017019660.1 XP_016875149.1 1 451
          XM_017019661.1 XP_016875150.1 2 165
          XM_017019662.1 XP_016875151.1 2 295
          XM_017019663.1 XP_016875152.1 1 451
          XM_017019664.1 XP_016875153.1 2 107
          XM_017019665.1 XP_016875154.1 1 451
          GenBank mRNA AF145204.1 - 9 1591
          AF164792.1 - 1 388
          AK289768.1 - 1 420
          AK294191.1 - 2 172
          AY281131.1 - 15 2473
          AY281132.1 - 1 148
          AY283057.2 - 8 1504
          AY620824.1 - 6 1198
          AY753193.1 - 1 148
          BC026313.1 - 1 350
          BC068451.1 - 2 111
          BC142669.1 - 1 402
          BC150204.1 - 1 402
          DA690060.1 - 1 455

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001271371, XM_006241248, XM_006241249, XM_008765239, XM_008765241, and others.
          This siRNA also targets the following transcripts in Mouse: NM_181398, NM_001177396, NM_001177398, NM_001128086, NM_001177397, and others.

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of synaptic plasticity by receptor localization to synapse
          ephrin receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0