Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173602.2 | NP_775873.2 | 3 | 445 | ||
XM_005269044.1 | XP_005269101.1 | 3 | 445 | |||
XM_006719520.3 | XP_006719583.1 | 3 | 391 | |||
XM_006719521.3 | XP_006719584.1 | 3 | 441 | |||
XM_011538594.2 | XP_011536896.1 | 3 | 441 | |||
XM_011538595.2 | XP_011536897.1 | 3 | 441 | |||
XM_011538596.2 | XP_011536898.1 | 3 | 441 | |||
XM_017019699.1 | XP_016875188.1 | 3 | 441 | |||
XM_017019700.1 | XP_016875189.1 | 3 | 445 | |||
XM_017019701.1 | XP_016875190.1 | 3 | 445 | |||
XM_017019702.1 | XP_016875191.1 | 3 | 445 | |||
XM_017019703.1 | XP_016875192.1 | 3 | 434 | |||
XM_017019704.1 | XP_016875193.1 | 3 | 445 | |||
XM_017019705.1 | XP_016875194.1 | 3 | 445 | |||
GenBank mRNA | AK091597.1 | - | 3 | 445 | ||
BC075027.2 | - | 3 | 323 | |||
BX537905.1 | - | 1 | 36 |