Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GOLGA8J SIRNA Assays ›
          Gene Symbol:
          GOLGA8J GOLGA8H GOLGA8IP GOLGA8K GOLGA8M GOLGA8N GOLGA8O GOLGA8T
          Gene Name:
          golgin A8 family member J
          golgin A8 family member H
          golgin A8 family member I, pseudogene
          golgin A8 family member K
          golgin A8 family member M
          golgin A8 family member N
          golgin A8 family member O
          golgin A8 family member T
          Gene Aliases:
          -
          GOLGA6L11
          GOLGA8I, GOLGA9P
          -
          -
          GOLGA8R
          -
          -
          Chromosome Location:
          Chr.15: 30082627 - 30096711 on Build GRCh38
          Chromosome Location:
          Chr.15: 30604030 - 30617827 on Build GRCh38
          Chromosome Location:
          Chr.15: 22610353 - 22617854 on Build GRCh38
          Chromosome Location:
          Chr.15: 32389585 - 32403292 on Build GRCh38
          Chromosome Location:
          Chr.15: 28698695 - 28718512 on Build GRCh38
          Chromosome Location:
          Chr.15: 32593456 - 32607310 on Build GRCh38
          Chromosome Location:
          Chr.15: 32441914 - 32456323 on Build GRCh38
          Chromosome Location:
          Chr.15: 30135787 - 30147192 on Build GRCh38
          UniGene ID:
          Hs.574257
          -
          Hs.375441
          -
          Hs.562625
          Hs.672754
          Hs.525905
          Hs.727141
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID 141714

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GOLGA8J
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282472.1 NP_001269401.1 11 916
          XM_006720651.3 XP_006720714.1 11 936
          XM_011521948.2 XP_011520250.1 11 1244
          XM_011521950.2 XP_011520252.1 11 1242
          Gene Symbol: GOLGA8H
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282490.1 NP_001269419.1 11 914
          Gene Symbol: GOLGA8IP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024074.1 10 1117
          GenBank mRNA AK093104.1 - 10 1117
          Gene Symbol: GOLGA8K
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282493.1 NP_001269422.1 11 916
          Gene Symbol: GOLGA8M
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282468.1 NP_001269397.1 11 916
          XM_005268284.4 XP_005268341.1 11 1715
          XM_005268285.4 XP_005268342.1 11 1715
          XM_006720654.3 XP_006720717.1 11 859
          XM_011521951.2 XP_011520253.1 12 1284
          XM_011521952.2 XP_011520254.1 11 1715
          XM_011521953.2 XP_011520255.1 11 1715
          XM_011521954.2 XP_011520256.1 11 849
          GenBank mRNA BF087405.1 - 3 188
          Gene Symbol: GOLGA8N
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282494.1 NP_001269423.1 11 916
          Gene Symbol: GOLGA8O
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001277308.1 NP_001264237.1 11 916
          XM_011521988.2 XP_011520290.1 11 1601
          XM_011521989.2 XP_011520291.1 11 1605
          GenBank mRNA BQ477899.1 - 6 446
          Gene Symbol: GOLGA8T
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_033933.1 11 936

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Process(es)

          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0