Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307955.1 | NP_001294884.1 | 3 | 359 | ||
NM_173629.2 | NP_775900.1 | 3 | 472 | |||
XM_011525923.2 | XP_011524225.1 | 5 | 562 | |||
XM_011525924.2 | XP_011524226.1 | 5 | 590 | |||
XM_011525925.2 | XP_011524227.1 | 3 | 567 | |||
XM_017025709.1 | XP_016881198.1 | 4 | 397 | |||
GenBank mRNA | AK096425.1 | - | 3 | 472 |