Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DPY19L2 SIRNA Assays ›
          Gene Symbol:
          DPY19L2 DPY19L2P1
          Gene Name:
          dpy-19 like 2
          DPY19L2 pseudogene 1
          Gene Aliases:
          SPATA34, SPGF9
          -
          Chromosome Location:
          Chr.12: 63558913 - 63669201 on Build GRCh38
          Chromosome Location:
          Chr.7: 35081287 - 35186162 on Build GRCh38
          UniGene ID:
          Hs.533644
          Hs.594815
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 141752

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DPY19L2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_173812.4 NP_776173.3 22 2408
          XM_006719352.2 XP_006719415.1 22 2334
          XM_011538215.2 XP_011536517.1 20 1874
          XM_011538218.2 XP_011536520.1 15 1629
          XM_017019188.1 XP_016874677.1 22 2455
          XM_017019189.1 XP_016874678.1 21 2395
          XM_017019190.1 XP_016874679.1 21 2329
          XM_017019191.1 XP_016874680.1 20 2305
          XM_017019192.1 XP_016874681.1 20 2293
          XM_017019193.1 XP_016874682.1 21 1990
          XM_017019194.1 XP_016874683.1 21 2017
          XM_017019195.1 XP_016874684.1 22 2065
          XM_017019196.1 XP_016874685.1 20 2154
          XM_017019197.1 XP_016874686.1 21 2288
          XM_017019198.1 XP_016874687.1 18 2009
          XM_017019199.1 XP_016874688.1 18 2078
          XM_017019201.1 XP_016874690.1 21 2251
          XM_017019202.1 XP_016874691.1 20 1864
          XM_017019203.1 XP_016874692.1 20 2086
          XM_017019204.1 XP_016874693.1 21 2324
          XM_017019205.1 XP_016874694.1 18 2178
          GenBank mRNA AK057511.1 - 22 2379
          AK303727.1 - 6 869
          AL833344.1 - 22 2335
          AY358792.1 - 22 2329
          BC031225.1 - 16 1410
          BC068442.1 - 22 2411 5297524
          Gene Symbol: DPY19L2P1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002833.2 22 2461
          GenBank mRNA BC066987.1 - 22 2461
          BX648666.1 - 18 1696

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          spermatid development
          protein C-linked glycosylation via 2'-alpha-mannosyl-L-tryptophan
          mannosyltransferase activity
          transferase activity, transferring glycosyl groups

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0