Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267774.1 | NP_001254703.1 | 6 | 734 | ||
NM_001267775.1 | NP_001254704.1 | 6 | 533 | |||
NM_001267776.1 | NP_001254705.1 | 5 | 497 | |||
NM_001267778.1 | NP_001254707.1 | 4 | 393 | |||
NM_174887.3 | NP_777547.1 | 6 | 615 | |||
GenBank mRNA | AA490722.1 | - | 4 | 309 | ||
AF070643.1 | - | 1 | 1105 | |||
AY224601.1 | - | 6 | 683 | |||
BC002640.2 | - | 6 | 595 | 3609552 | ||
BC038094.1 | - | 5 | 463 | 3907361 |