Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFIKKN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175575.5 | NP_783165.1 | 2 | 2539 | ||
XM_017024172.1 | XP_016879661.1 | 2 | 2018 | |||
GenBank mRNA | AK127743.1 | - | 2 | 2199 | ||
AK131196.1 | - | 1109 | ||||
AK298172.1 | - | 2 | 2018 | |||
AY358142.1 | - | 2 | 2374 | |||
BC140842.1 | - | 2 | 1933 | 9021710 | ||
BC140844.1 | - | 2 | 1933 | 9021712 |