Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ERC1 SIRNA Assays ›
Gene Symbol:
ERC1
Gene Name:
ELKS/RAB6-interacting/CAST family member 1
Gene Aliases:
Cast2, ELKS, ERC-1, RAB6IP2
Chromosome Location:
Chr.12: 990010 - 1495933 on Build GRCh38
UniGene ID:
Hs.601216
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
141812
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ERC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001301248.1
NP_001288177.1
1
529
NM_178039.3
NP_829883.1
2
644
NM_178040.3
NP_829884.1
2
644
NR_027946.2
2
809
NR_027948.2
2
809
NR_027949.2
2
644
XM_011520941.2
XP_011519243.1
2
688
XM_017019052.1
XP_016874541.1
2
754
XM_017019053.1
XP_016874542.1
2
688
XM_017019054.1
XP_016874543.1
2
614
XM_017019055.1
XP_016874544.1
2
610
XM_017019056.1
XP_016874545.1
2
551
XM_017019057.1
XP_016874546.1
2
754
XM_017019058.1
XP_016874547.1
2
688
XM_017019059.1
XP_016874548.1
2
754
XM_017019060.1
XP_016874549.1
2
754
XM_017019061.1
XP_016874550.1
2
754
XM_017019062.1
XP_016874551.1
2
754
XM_017019063.1
XP_016874552.1
2
754
XM_017019064.1
XP_016874553.1
2
754
XM_017019065.1
XP_016874554.1
2
754
XM_017019066.1
XP_016874555.1
2
754
XM_017019067.1
XP_016874556.1
2
754
XM_017019068.1
XP_016874557.1
2
754
XM_017019069.1
XP_016874558.1
2
754
XM_017019070.1
XP_016874559.1
2
754
XM_017019071.1
XP_016874560.1
2
754
XM_017019072.1
XP_016874561.1
2
754
XM_017019073.1
XP_016874562.1
2
754
GenBank mRNA
AB015617.1
-
2
767
AB029004.2
-
1
496
AB053468.1
-
2
750
AB053469.1
-
2
579
AB053470.1
-
2
586
AB053471.1
-
2
554
AK091577.1
-
2
754
BC068006.1
-
1
495
BC132782.1
-
1
505
40146804
BC132784.1
-
1
505
40146806
BC144287.1
-
1
505
BC150248.1
-
1
517
9007473
DR000847.1
-
2
667
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
I-kappaB phosphorylation
multicellular organism development
protein transport
retrograde transport, endosome to Golgi
positive regulation of NF-kappaB transcription factor activity
cell-cell adhesion
protein binding
Rab GTPase binding
PDZ domain binding
leucine zipper domain binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0