Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178836.3 | NP_849158.2 | 2 | 2088 | ||
XM_017024310.1 | XP_016879799.1 | 2 | 2084 | |||
GenBank mRNA | AK090899.1 | - | 2 | 2065 | ||
AK098419.1 | - | 1 | 1207 | |||
BC031263.1 | - | 2 | 2074 | 5274952 |