Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATXN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000333.3 | NP_000324.1 | 6 | 1189 | ||
NM_001128149.2 | NP_001121621.2 | 2 | 369 | |||
NM_001177387.1 | NP_001170858.1 | 5 | 746 | |||
GenBank mRNA | AF032103.1 | - | 4 | 328 | ||
AF032105.1 | - | 6 | 1189 | |||
AJ000517.1 | - | 6 | 1197 | |||
AK304062.1 | - | 2 | 292 | |||
AK310745.1 | - | 2 | 306 | |||
BP220889.1 | - | 2 | 370 |