Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000582.2 | NP_000573.1 | 6 | 719 | ||
NM_001040058.1 | NP_001035147.1 | 7 | 761 | |||
NM_001040060.1 | NP_001035149.1 | 6 | 680 | |||
NM_001251829.1 | NP_001238758.1 | 5 | 638 | |||
NM_001251830.1 | NP_001238759.1 | 8 | 950 | |||
XM_017008564.1 | XP_016864053.1 | 5 | 1187 | |||
GenBank mRNA | AB209987.1 | - | 4 | 473 | ||
AB469789.1 | - | 4 | 473 | |||
AF052124.1 | - | 6 | 641 | |||
AK057738.1 | - | 7 | 697 | |||
AK075463.1 | - | 8 | 631 | |||
AK290090.1 | - | 6 | 616 | |||
AK290104.1 | - | 7 | 698 | |||
AK295491.1 | - | 8 | 885 | |||
AK296035.1 | - | 6 | 616 | |||
AK315461.1 | - | 6 | 649 | |||
BC007016.1 | - | 6 | 655 | 4052438 | ||
BC017387.1 | - | 7 | 701 | 3828885 | ||
BC022844.1 | - | 6 | 656 | 4284921 | ||
BC093033.1 | - | 6 | 672 | 5288027 | ||
BX648003.1 | - | 6 | 634 | |||
D28759.1 | - | 6 | 596 | |||
D28760.1 | - | 5 | 554 | |||
D28761.1 | - | 5 | 515 | |||
DQ839491.1 | - | 6 | 596 | |||
DQ846870.1 | - | 6 | 596 | |||
DQ846871.1 | - | 6 | 596 | |||
J04765.1 | - | 6 | 662 | |||
JF412666.1 | - | 7 | 532 | |||
JF412667.1 | - | 6 | 678 | |||
KR062183.1 | - | 8 | 885 | |||
M83248.1 | - | 7 | 693 | |||
X13694.1 | - | 7 | 663 |