Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RARG SIRNA Assays ›
          Gene Symbol:
          RARG
          Gene Name:
          retinoic acid receptor gamma
          Gene Aliases:
          NR1B3, RARC
          Chromosome Location:
          Chr.12: 53210566 - 53232256 on Build GRCh38
          UniGene ID:
          Hs.1497
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 142162

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RARG
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000966.5 NP_000957.1 6 1048
          NM_001042728.2 NP_001036193.1 4 836
          NM_001243730.1 NP_001230659.1 5 722
          NM_001243731.1 NP_001230660.1 4 573
          NM_001243732.1 NP_001230661.1 3 687
          GenBank mRNA AB307689.1 - 4 556
          AK290588.1 - 6 1044
          AK297023.1 - 5 640
          AK297110.1 - 4 549
          AK297277.1 - 3 687
          AK297801.1 - 4 682
          AK301057.1 - 4 831
          AK316259.1 - 4 560
          BC093727.1 - 4 580 7939572
          BC093729.1 - 4 580 7939574
          HQ692828.1 - 4 556
          HQ692829.1 - 4 523
          L12060.1 - 5 664
          M24857.1 - 6 970
          M38258.1 - 4 702
          M57707.1 - 5 755

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          neural tube closure
          glandular epithelial cell development
          retinal pigment epithelium development
          growth plate cartilage chondrocyte growth
          transcription initiation from RNA polymerase II promoter
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          regulation of cell size
          anterior/posterior pattern specification
          embryonic camera-type eye development
          regulation of myelination
          negative regulation of chondrocyte differentiation
          response to retinoic acid
          embryonic hindlimb morphogenesis
          multicellular organism growth
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of programmed cell death
          steroid hormone mediated signaling pathway
          regulation of myeloid cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          embryonic eye morphogenesis
          retinoic acid receptor signaling pathway
          canonical Wnt signaling pathway
          face development
          trachea cartilage development
          prostate gland epithelium morphogenesis
          Harderian gland development
          cellular response to retinoic acid
          RNA polymerase II regulatory region sequence-specific DNA binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          retinoic acid receptor activity
          protein binding
          zinc ion binding
          retinoid X receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0