Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002751.6 | NP_002742.3 | 4 | 484 | ||
NR_110887.1 | 4 | 484 | ||||
XM_011530706.1 | XP_011529008.1 | 2 | 208 | |||
GenBank mRNA | AF001008.1 | - | 4 | 383 | ||
AF001174.1 | - | 4 | 389 | |||
AF031135.1 | - | 4 | 398 | |||
AK291845.1 | - | 4 | 435 | |||
AK299745.1 | - | 3 | 396 | |||
AK308969.1 | - | 4 | 400 | |||
AK309046.1 | - | 4 | 586 | |||
BC027933.1 | - | 4 | 441 | 5199628 | ||
BX361670.2 | - | 4 | 468 | |||
BX444192.2 | - | 4 | 483 | |||
CR456514.1 | - | 4 | 385 | |||
U53442.1 | - | 4 | 426 | |||
U92268.1 | - | 4 | 387 | |||
Y14440.1 | - | 4 | 389 |