Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RAP1GAP SIRNA Assays ›
Gene Symbol:
RAP1GAP
Gene Name:
RAP1 GTPase activating protein
Gene Aliases:
RAP1GA1, RAP1GAP1, RAP1GAPII, RAPGAP
Chromosome Location:
Chr.1: 21596215 - 21669444 on Build GRCh38
UniGene ID:
Hs.148178
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
142444
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RAP1GAP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145657.1
NP_001139129.1
8, 9
667
NM_001145658.1
NP_001139130.1
7, 8
527
NM_002885.2
NP_002876.2
7, 8
560
XM_005245955.3
XP_005246012.2
7, 8
572
XM_006710804.2
XP_006710867.2
7, 8
572
XM_006710805.2
XP_006710868.2
7, 8
572
XM_017001965.1
XP_016857454.1
6, 7
574
XM_017001966.1
XP_016857455.1
6, 7
574
XM_017001967.1
XP_016857456.1
7, 8
572
XM_017001968.1
XP_016857457.1
7, 8
607
XM_017001969.1
XP_016857458.1
6, 7
574
XM_017001970.1
XP_016857459.1
6, 7
574
XM_017001971.1
XP_016857460.1
7, 8
526
XM_017001972.1
XP_016857461.1
7, 8
572
XM_017001973.1
XP_016857462.1
6, 7
605
XM_017001974.1
XP_016857463.1
7, 8
572
XM_017001975.1
XP_016857464.1
6, 7
574
XM_017001976.1
XP_016857465.1
7, 8
572
XM_017001977.1
XP_016857466.1
7, 8
572
XM_017001978.1
XP_016857467.1
8, 9
721
XM_017001979.1
XP_016857468.1
9, 10
725
XM_017001980.1
XP_016857469.1
7, 8
526
XM_017001981.1
XP_016857470.1
9, 10
701
XM_017001982.1
XP_016857471.1
8, 9
721
XM_017001983.1
XP_016857472.1
10, 11
933
XM_017001984.1
XP_016857473.1
9, 10
701
XM_017001985.1
XP_016857474.1
7, 8
526
XM_017001986.1
XP_016857475.1
9, 10
738
XM_017001987.1
XP_016857476.1
8, 9
667
XM_017001988.1
XP_016857477.1
8, 9
638
XM_017001989.1
XP_016857478.1
9, 10
738
XM_017001990.1
XP_016857479.1
8, 9
623
XM_017001991.1
XP_016857480.1
8, 9
661
GenBank mRNA
AB007943.3
-
8, 9
605
BC035030.1
-
4, 5
316
5261027
BC054490.1
-
7, 8
467
5767775
BP283134.1
-
7, 8
560
CV574545.1
-
7, 8
517
M64788.1
-
7, 8
503
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
regulation of GTPase activity
positive regulation of GTPase activity
negative regulation of neuron differentiation
regulation of small GTPase mediated signal transduction
negative regulation of microvillus assembly
negative regulation of GTP binding
negative regulation of thyroid gland epithelial cell proliferation
cellular response to glial cell derived neurotrophic factor
GTPase activity
GTPase activator activity
protein binding
Ras GTPase binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-zgjsk:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0