Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270427.1 | NP_001257356.1 | 2 | 340 | ||
NM_005047.3 | NP_005038.1 | 2 | 340 | |||
XM_011518866.2 | XP_011517168.1 | 2 | 250 | |||
XM_017014927.1 | XP_016870416.1 | 2 | 250 | |||
XM_017014928.1 | XP_016870417.1 | 2 | 250 | |||
GenBank mRNA | AK291051.1 | - | 2 | 241 | ||
AK303007.1 | - | 2 | 270 | |||
BC014478.1 | - | 2 | 229 | 4878087 | ||
BM835267.1 | - | 2 | 247 | |||
D31889.1 | - | 2 | 227 | |||
HY023459.1 | - | 2 | 341 | |||
S79862.1 | - | 2 | 231 |