Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004161.4 | NP_004152.1 | 5, 6 | 814 | ||
XM_005264468.1 | XP_005264525.1 | 4, 5 | 523 | |||
GenBank mRNA | AF498929.1 | - | 5, 6 | 428 | ||
AK055927.1 | - | 3, 4 | 429 | |||
AK303678.1 | - | 4, 5 | 522 | |||
AK315495.1 | - | 5, 6 | 623 | |||
BC000905.2 | - | 5, 6 | 606 | 2900705 | ||
BT019528.1 | - | 5, 6 | 428 | |||
BT019529.1 | - | 5, 6 | 428 | |||
BX571747.1 | - | 5, 6 | 611 | |||
CR536488.1 | - | 5, 6 | 428 | |||
M28209.1 | - | 5, 6 | 478 | |||
X13898.1 | - | 5, 6 | 428 |