Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTPN6 SIRNA Assays ›
          Gene Symbol:
          PTPN6
          Gene Name:
          protein tyrosine phosphatase, non-receptor type 6
          Gene Aliases:
          HCP, HCPH, HPTP1C, PTP-1C, SH-PTP1, SHP-1, SHP-1L, SHP1
          Chromosome Location:
          Chr.12: 6946576 - 6961316 on Build GRCh38
          UniGene ID:
          Hs.63489
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 143142

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTPN6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002831.5 NP_002822.2 3 399
          NM_080548.4 NP_536858.1 3 312
          NM_080549.3 NP_536859.1 3 399
          XM_006718994.1 XP_006719057.1 3 456
          XM_011520988.1 XP_011519290.1 3 313
          GenBank mRNA AF178946.1 - 3 181
          AK223629.1 - 3 294
          AK290421.1 - 3 282
          AK298470.1 - 2 565
          AK300167.1 - 2 156
          AK308230.1 - 3 347
          AK309037.1 - 2 511
          AK313379.1 - 3 312
          BC002523.2 - 3 298 3140265
          BC007667.2 - 3 298 3138804
          BG754792.1 - 3 332
          BM742181.1 - 3 312
          BT007390.1 - 2 163
          CR986427.1 - 3 390
          DA431502.1 - 3 312
          DA868001.1 - 3 316
          DC297194.1 - 3 399
          M74903.1 - 3 240
          M77273.1 - 3 301

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          hematopoietic progenitor cell differentiation
          negative regulation of humoral immune response mediated by circulating immunoglobulin
          protein dephosphorylation
          apoptotic process
          G-protein coupled receptor signaling pathway
          cell proliferation
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          positive regulation of phosphatidylinositol 3-kinase signaling
          peptidyl-tyrosine phosphorylation
          cell differentiation
          platelet activation
          platelet formation
          T cell costimulation
          abortive mitotic cell cycle
          positive regulation of cell adhesion mediated by integrin
          peptidyl-tyrosine dephosphorylation
          intracellular signal transduction
          megakaryocyte development
          negative regulation of T cell proliferation
          natural killer cell mediated cytotoxicity
          negative regulation of MAP kinase activity
          regulation of B cell differentiation
          negative regulation of peptidyl-tyrosine phosphorylation
          B cell receptor signaling pathway
          negative regulation of B cell receptor signaling pathway
          negative regulation of T cell receptor signaling pathway
          leukocyte migration
          regulation of release of sequestered calcium ion into cytosol
          regulation of type I interferon-mediated signaling pathway
          regulation of ERK1 and ERK2 cascade
          platelet aggregation
          regulation of G1/S transition of mitotic cell cycle
          phosphotyrosine binding
          protein tyrosine phosphatase activity
          transmembrane receptor protein tyrosine phosphatase activity
          protein binding
          SH3 domain binding
          protein kinase binding
          SH2 domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0