Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other OPRM1 SIRNA Assays ›
          Gene Symbol:
          OPRM1
          Gene Name:
          opioid receptor mu 1
          Gene Aliases:
          LMOR, M-OR-1, MOP, MOR, MOR1, OPRM
          Chromosome Location:
          Chr.6: 154010496 - 154246867 on Build GRCh38
          UniGene ID:
          Hs.2353
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 143366

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: OPRM1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000914.4 NP_000905.3 2, 3 1094
          NM_001008503.2 NP_001008503.2 2, 3 785
          NM_001008504.3 NP_001008504.2 2, 3 1094
          NM_001008505.2 NP_001008505.2 2, 3 785
          NM_001145279.3 NP_001138751.1 4, 5 1453
          NM_001145280.3 NP_001138752.1 2, 3 884
          NM_001145281.2 NP_001138753.1 2, 3 425
          NM_001145282.2 NP_001138754.1 2, 3 785
          NM_001145283.2 NP_001138755.1 2, 3 785
          NM_001145284.3 NP_001138756.1 2, 3 785
          NM_001145285.2 NP_001138757.1 2, 3 785
          NM_001145286.2 NP_001138758.1 2, 3 785
          NM_001145287.2 NP_001138759.1 2, 3 1659
          NM_001285523.1 NP_001272452.1 2, 3 1094
          NM_001285524.1 NP_001272453.1 3, 4 989
          NM_001285526.1 NP_001272455.1 2, 3 1640
          NM_001285527.1 NP_001272456.1 1, 2 425
          NM_001285528.1 NP_001272457.1 1, 2 425
          NR_104348.1 2, 3 785
          NR_104349.1 2, 3 785
          NR_104351.1 2, 3 785
          XM_011535851.2 XP_011534153.1 3, 4 1575
          XM_011535853.2 XP_011534155.1 3, 4 616
          XM_011535856.2 XP_011534158.1 2, 3 448
          XM_011535862.2 XP_011534164.1 2, 3 544
          XM_017010903.1 XP_016866392.1 2, 3 2569
          XM_017010904.1 XP_016866393.1 2, 3 1678
          XM_017010905.1 XP_016866394.1 2, 3 1697
          XM_017010906.1 XP_016866395.1 2, 3 2569
          GenBank mRNA AK313901.1 - 2, 3 834
          AY036622.1 - 2, 3 700
          AY036623.1 - 2, 3 700
          AY195733.2 - 1, 2 371
          AY225404.1 - 2, 3 742
          AY309001.1 - 2, 3 701
          AY309005.1 - 2, 3 701
          AY309006.1 - 2, 3 701
          AY309007.1 - 2, 3 701
          AY309008.1 - 2, 3 701
          AY309009.1 - 2, 3 701
          AY364230.1 - 2, 3 721
          AY364890.1 - 1, 2 425
          AY521028.1 - 2, 3 651
          BC074927.2 - 2, 3 696 30915262
          EU340241.1 - 4, 5 1448
          EU340242.1 - 2, 3 879
          EU340243.1 - 2, 3 420
          EU360599.2 - 2, 3 1640
          EU362990.2 - 2, 3 1659
          FJ041291.1 - 2, 3 1094
          FJ041292.1 - 3, 4 989
          FJ041293.1 - 2, 3 699
          FJ041294.1 - 2, 3 699
          GQ258059.1 - 2, 3 583
          JX914655.1 - 2, 3 888
          L25119.1 - 2, 3 863
          L29301.1 - 2, 3 889
          U12569.1 - 2, 3 847

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          acute inflammatory response to antigenic stimulus
          G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
          adenylate cyclase-activating dopamine receptor signaling pathway
          negative regulation of adenylate cyclase activity
          phospholipase C-activating G-protein coupled receptor signaling pathway
          neuropeptide signaling pathway
          chemical synaptic transmission
          sensory perception
          locomotory behavior
          negative regulation of cell proliferation
          response to radiation
          sensory perception of pain
          adenylate cyclase-inhibiting opioid receptor signaling pathway
          response to food
          positive regulation of appetite
          response to lipopolysaccharide
          cellular response to stress
          wound healing
          response to cocaine
          eating behavior
          estrous cycle
          behavioral response to ethanol
          positive regulation of neurogenesis
          regulation of sensory perception of pain
          excitatory postsynaptic potential
          negative regulation of Wnt protein secretion
          positive regulation of ERK1 and ERK2 cascade
          calcium ion transmembrane transport
          response to growth factor
          cellular response to morphine
          regulation of N-methyl-D-aspartate selective glutamate receptor activity
          G-protein alpha-subunit binding
          G-protein coupled receptor activity
          beta-endorphin receptor activity
          voltage-gated calcium channel activity
          protein binding
          protein C-terminus binding
          protein domain specific binding
          filamin binding
          G-protein beta-subunit binding
          morphine receptor activity
          neuropeptide binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0