Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRIP12 SIRNA Assays ›
Gene Symbol:
TRIP12
Gene Name:
thyroid hormone receptor interactor 12
Gene Aliases:
TRIP-12, ULF
Chromosome Location:
Chr.2: 229763837 - 229923234 on Build GRCh38
UniGene ID:
Hs.572642
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
14339
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRIP12
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001284214.1
NP_001271143.1
4
454
NM_001284215.1
NP_001271144.1
2
198
NM_004238.2
NP_004229.1
3
328
XM_005246954.4
XP_005247011.1
4
478
XM_005246955.4
XP_005247012.1
4
477
XM_005246956.4
XP_005247013.1
4
457
XM_005246957.4
XP_005247014.1
4
444
XM_005246958.4
XP_005247015.1
4
478
XM_005246960.4
XP_005247017.1
4
478
XM_005246961.3
XP_005247018.1
3
328
XM_005246962.4
XP_005247019.1
3
328
XM_006712852.3
XP_006712915.1
3
281
XM_011512180.2
XP_011510482.1
4
431
XM_011512181.2
XP_011510483.1
4
429
XM_011512183.2
XP_011510485.1
3
285
XM_017005272.1
XP_016860761.1
5
537
XM_017005273.1
XP_016860762.1
4
404
XM_017005274.1
XP_016860763.1
4
352
XM_017005275.1
XP_016860764.1
4
410
XM_017005276.1
XP_016860765.1
4
415
XM_017005277.1
XP_016860766.1
5
555
XM_017005278.1
XP_016860767.1
4
370
XM_017005279.1
XP_016860768.1
4
442
XM_017005280.1
XP_016860769.1
4
431
XM_017005281.1
XP_016860770.1
4
431
XM_017005282.1
XP_016860771.1
4
477
XM_017005283.1
XP_016860772.1
4
478
XM_017005284.1
XP_016860773.1
4
478
XM_017005285.1
XP_016860774.1
3
315
XM_017005286.1
XP_016860775.1
3
350
XM_017005287.1
XP_016860776.1
3
331
XM_017005288.1
XP_016860777.1
3
293
XM_017005289.1
XP_016860778.1
4
478
XM_017005290.1
XP_016860779.1
4
478
XM_017005291.1
XP_016860780.1
3
328
XM_017005292.1
XP_016860781.1
3
328
XM_017005293.1
XP_016860782.1
3
328
GenBank mRNA
AK302468.1
-
4
408
AK310971.1
-
3
328
BC114556.1
-
4
348
40083165
D28476.1
-
3
258
DB240964.1
-
3
328
EU489742.1
-
2
152
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001031659, XM_008767252, XM_008767257, XM_006245379, XM_006245377, and others.This siRNA also targets the following transcripts in Mouse: NM_133975, AK158807, AK134397, AK164719.
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
DNA repair
cellular response to DNA damage stimulus
embryo development
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of histone H2A K63-linked ubiquitination
negative regulation of double-strand break repair
ubiquitin-protein transferase activity
protein binding
ligase activity
thyroid hormone receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65b4bdbffd-zctgw:80/10.239.163.131:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0