Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ABLIM1 SIRNA Assays ›
          Gene Symbol:
          ABLIM1
          Gene Name:
          actin binding LIM protein 1
          Gene Aliases:
          ABLIM, LIMAB1, LIMATIN, abLIM-1
          Chromosome Location:
          Chr.10: 114431110 - 114821643 on Build GRCh38
          UniGene ID:
          Hs.438236
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 143452

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ABLIM1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001003407.1 NP_001003407.1 10 1282
          NM_001003408.1 NP_001003408.1 11 1366
          NM_001322882.1 NP_001309811.1 11 1366
          NM_001322883.1 NP_001309812.1 10 1282
          NM_001322884.1 NP_001309813.1 10 1081
          NM_001322885.1 NP_001309814.1 10 1081
          NM_001322886.1 NP_001309815.1 10 1081
          NM_001322887.1 NP_001309816.1 10 1282
          NM_001322888.1 NP_001309817.1 5 533
          NM_001322889.1 NP_001309818.1 6 638
          NM_001322890.1 NP_001309819.1 5 533
          NM_001322891.1 NP_001309820.1 5 554
          NM_001322892.1 NP_001309821.1 5 533
          NM_001322893.1 NP_001309822.1 5 554
          NM_001322894.1 NP_001309823.1 5 554
          NM_001322895.1 NP_001309824.1 5 533
          NM_001322896.1 NP_001309825.1 4 455
          NM_001322897.1 NP_001309826.1 4 455
          NM_001322898.1 NP_001309827.1 5 533
          NM_001322899.1 NP_001309828.1 5 554
          NM_001322900.1 NP_001309829.1 4 455
          NM_002313.5 NP_002304.3 10 1259
          NM_006720.3 NP_006711.3 6 617
          XM_005269824.2 XP_005269881.1 10 2018
          XM_005269826.1 XP_005269883.1 6 622
          XM_005269827.4 XP_005269884.1 5 1252
          XM_006717837.1 XP_006717900.1 11 1714
          XM_006717846.3 XP_006717909.1 6 1357
          XM_006717847.1 XP_006717910.1 5 451
          XM_011539801.1 XP_011538103.1 11 1118
          XM_011539802.1 XP_011538104.1 11 1031
          XM_011539803.1 XP_011538105.1 12 1136
          XM_011539805.1 XP_011538107.1 11 1371
          XM_017016242.1 XP_016871731.1 13 1220
          XM_017016243.1 XP_016871732.1 13 1220
          XM_017016244.1 XP_016871733.1 13 1220
          XM_017016245.1 XP_016871734.1 11 1204
          XM_017016246.1 XP_016871735.1 13 1220
          XM_017016247.1 XP_016871736.1 10 1000
          XM_017016248.1 XP_016871737.1 9 916
          XM_017016249.1 XP_016871738.1 13 1220
          XM_017016250.1 XP_016871739.1 10 1120
          XM_017016251.1 XP_016871740.1 12 1136
          XM_017016252.1 XP_016871741.1 12 1136
          XM_017016253.1 XP_016871742.1 12 1136
          XM_017016254.1 XP_016871743.1 13 1220
          XM_017016255.1 XP_016871744.1 10 1120
          XM_017016256.1 XP_016871745.1 10 2018
          XM_017016257.1 XP_016871746.1 12 1220
          XM_017016258.1 XP_016871747.1 5 1252
          XM_017016259.1 XP_016871748.1 5 451
          GenBank mRNA AF005654.1 - 10 1280
          AK093471.1 - 11 886
          AK098277.1 - 6 622
          AK122891.1 - 11 1716
          AK297344.1 - 10 1356
          AK298709.1 - 5 437
          BC002448.2 - 4 359 3347450
          CR749819.1 - 5 529
          D31883.2 - 10 1084

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cytoskeleton organization
          visual perception
          organ morphogenesis
          lamellipodium assembly
          cilium assembly
          actin binding
          protein binding
          zinc ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-9c8cf:80/100.66.75.150:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0