Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABLIM1 SIRNA Assays ›
Gene Symbol:
ABLIM1
Gene Name:
actin binding LIM protein 1
Gene Aliases:
ABLIM, LIMAB1, LIMATIN, abLIM-1
Chromosome Location:
Chr.10: 114431110 - 114821643 on Build GRCh38
UniGene ID:
Hs.438236
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
143453
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABLIM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001003407.1
NP_001003407.1
23
2404
NM_001003408.1
NP_001003408.1
24
2488
NM_001322882.1
NP_001309811.1
24
2494
NM_001322883.1
NP_001309812.1
22
2299
NM_001322884.1
NP_001309813.1
22
2104
NM_001322885.1
NP_001309814.1
22
2098
NM_001322886.1
NP_001309815.1
22
2062
NM_001322887.1
NP_001309816.1
21
2158
NM_001322888.1
NP_001309817.1
18
1634
NM_001322889.1
NP_001309818.1
18
1619
NM_001322890.1
NP_001309819.1
17
1556
NM_001322891.1
NP_001309820.1
17
1577
NM_001322892.1
NP_001309821.1
17
1550
NM_001322893.1
NP_001309822.1
17
1571
NM_001322894.1
NP_001309823.1
17
1535
NM_001322895.1
NP_001309824.1
17
1514
NM_001322896.1
NP_001309825.1
16
1472
NM_001322897.1
NP_001309826.1
16
1436
NM_001322898.1
NP_001309827.1
16
1409
NM_001322899.1
NP_001309828.1
16
1430
NM_001322900.1
NP_001309829.1
15
1331
NM_002313.5
NP_002304.3
23
2381
NM_006720.3
NP_006711.3
18
1634
XM_005269824.2
XP_005269881.1
22
2999
XM_005269826.1
XP_005269883.1
20
1870
XM_005269827.4
XP_005269884.1
19
2500
XM_006717837.1
XP_006717900.1
25
2962
XM_006717846.3
XP_006717909.1
20
2605
XM_006717847.1
XP_006717910.1
19
1699
XM_011539801.1
XP_011538103.1
25
2366
XM_011539802.1
XP_011538104.1
25
2279
XM_011539803.1
XP_011538105.1
25
2258
XM_011539805.1
XP_011538107.1
25
2619
XM_017016242.1
XP_016871731.1
27
2462
XM_017016243.1
XP_016871732.1
26
2348
XM_017016244.1
XP_016871733.1
26
2342
XM_017016245.1
XP_016871734.1
25
2452
XM_017016246.1
XP_016871735.1
25
2243
XM_017016247.1
XP_016871736.1
22
2017
XM_017016248.1
XP_016871737.1
22
2017
XM_017016249.1
XP_016871738.1
25
2201
XM_017016250.1
XP_016871739.1
24
2368
XM_017016251.1
XP_016871740.1
24
2159
XM_017016252.1
XP_016871741.1
24
2153
XM_017016253.1
XP_016871742.1
24
2117
XM_017016254.1
XP_016871743.1
24
2096
XM_017016255.1
XP_016871744.1
23
2248
XM_017016256.1
XP_016871745.1
22
3041
XM_017016258.1
XP_016871747.1
18
2380
XM_017016259.1
XP_016871748.1
18
1579
GenBank mRNA
AF005654.1
-
23
2402
AK058007.1
-
13
1023
AK093471.1
-
23
1903
AK098277.1
-
18
1639
AK122891.1
-
24
2844
AK297344.1
-
22
2337
AK298709.1
-
16
1313
BC002448.2
-
16
1376
3347450
CR749819.1
-
17
1546
D31883.2
-
23
2206
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
visual perception
organ morphogenesis
lamellipodium assembly
cilium assembly
actin binding
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-f6n66:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0