Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ABLIM1 SIRNA Assays ›
Gene Symbol:
ABLIM1
Gene Name:
actin binding LIM protein 1
Gene Aliases:
ABLIM, LIMAB1, LIMATIN, abLIM-1
Chromosome Location:
Chr.10: 114431110 - 114821643 on Build GRCh38
UniGene ID:
Hs.438236
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
143454
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ABLIM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001003407.1
NP_001003407.1
23
2453
NM_001003408.1
NP_001003408.1
24
2537
NM_001322882.1
NP_001309811.1
24
2543
NM_001322883.1
NP_001309812.1
22
2348
NM_001322884.1
NP_001309813.1
22
2153
NM_001322885.1
NP_001309814.1
22
2147
NM_001322886.1
NP_001309815.1
22
2111
NM_001322887.1
NP_001309816.1
21
2207
NM_001322888.1
NP_001309817.1
18
1683
NM_001322889.1
NP_001309818.1
18
1668
NM_001322890.1
NP_001309819.1
17
1605
NM_001322891.1
NP_001309820.1
17
1626
NM_001322892.1
NP_001309821.1
17
1599
NM_001322893.1
NP_001309822.1
17
1620
NM_001322894.1
NP_001309823.1
17
1584
NM_001322895.1
NP_001309824.1
17
1563
NM_001322896.1
NP_001309825.1
16
1521
NM_001322897.1
NP_001309826.1
16
1485
NM_001322898.1
NP_001309827.1
16
1458
NM_001322899.1
NP_001309828.1
16
1479
NM_001322900.1
NP_001309829.1
15
1380
NM_002313.5
NP_002304.3
23
2430
NM_006720.3
NP_006711.3
18
1683
XM_005269824.2
XP_005269881.1
22
3048
XM_005269826.1
XP_005269883.1
20
1919
XM_005269827.4
XP_005269884.1
19
2549
XM_006717837.1
XP_006717900.1
25
3011
XM_006717846.3
XP_006717909.1
20
2654
XM_006717847.1
XP_006717910.1
19
1748
XM_011539801.1
XP_011538103.1
25
2415
XM_011539802.1
XP_011538104.1
25
2328
XM_011539803.1
XP_011538105.1
25
2307
XM_011539805.1
XP_011538107.1
25
2668
XM_017016242.1
XP_016871731.1
27
2511
XM_017016243.1
XP_016871732.1
26
2397
XM_017016244.1
XP_016871733.1
26
2391
XM_017016245.1
XP_016871734.1
25
2501
XM_017016246.1
XP_016871735.1
25
2292
XM_017016247.1
XP_016871736.1
22
2066
XM_017016248.1
XP_016871737.1
22
2066
XM_017016249.1
XP_016871738.1
25
2250
XM_017016250.1
XP_016871739.1
24
2417
XM_017016251.1
XP_016871740.1
24
2208
XM_017016252.1
XP_016871741.1
24
2202
XM_017016253.1
XP_016871742.1
24
2166
XM_017016254.1
XP_016871743.1
24
2145
XM_017016255.1
XP_016871744.1
23
2297
XM_017016256.1
XP_016871745.1
22
3090
XM_017016258.1
XP_016871747.1
18
2429
XM_017016259.1
XP_016871748.1
18
1628
GenBank mRNA
AF005654.1
-
23
2451
AK058007.1
-
13
1072
AK093471.1
-
23
1952
AK098277.1
-
18
1688
AK122891.1
-
24
2893
AK297344.1
-
22
2386
AK298709.1
-
16
1362
BC002448.2
-
16
1425
3347450
CR749819.1
-
17
1595
D31883.2
-
23
2255
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
visual perception
organ morphogenesis
lamellipodium assembly
cilium assembly
actin binding
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-pwjgx:80/100.66.75.150:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0