Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LYN SIRNA Assays ›
          Gene Symbol:
          LYN
          Gene Name:
          LYN proto-oncogene, Src family tyrosine kinase
          Gene Aliases:
          JTK8, p53Lyn, p56Lyn
          Chromosome Location:
          Chr.8: 55877305 - 56012447 on Build GRCh38
          UniGene ID:
          Hs.491767
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 143467

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LYN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002350.3 NP_002341.1 2 357
          GenBank mRNA AK290494.1 - 2 267
          BC075001.2 - 1 100 30915399
          BC075002.2 - 1 100 30915611
          BC126456.1 - 3 194 8992172
          BC126458.1 - 3 241 8992174
          M16038.1 - 2 372

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          B cell homeostasis
          regulation of cytokine production
          regulation of protein phosphorylation
          negative regulation of protein phosphorylation
          stimulatory C-type lectin receptor signaling pathway
          adaptive immune response
          Fc receptor mediated stimulatory signaling pathway
          tolerance induction to self antigen
          histamine secretion by mast cell
          platelet degranulation
          negative regulation of myeloid leukocyte differentiation
          immune response-regulating cell surface receptor signaling pathway
          Fc receptor mediated inhibitory signaling pathway
          regulation of B cell apoptotic process
          protein phosphorylation
          inflammatory response
          cellular response to DNA damage stimulus
          response to sterol depletion
          signal transduction
          transmembrane receptor protein tyrosine kinase signaling pathway
          central nervous system development
          blood coagulation
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          response to toxic substance
          response to hormone
          response to carbohydrate
          positive regulation of neuron projection development
          oligodendrocyte development
          response to organic cyclic compound
          viral process
          peptidyl-tyrosine phosphorylation
          signal transduction by protein phosphorylation
          platelet activation
          erythrocyte differentiation
          positive regulation of cell migration
          negative regulation of B cell proliferation
          neuron projection development
          T cell costimulation
          lipopolysaccharide-mediated signaling pathway
          cellular response to extracellular stimulus
          response to insulin
          regulation of mast cell activation
          regulation of cell adhesion mediated by integrin
          negative regulation of toll-like receptor 2 signaling pathway
          negative regulation of toll-like receptor 4 signaling pathway
          cellular response to heat
          peptidyl-tyrosine autophosphorylation
          Fc-epsilon receptor signaling pathway
          Fc-gamma receptor signaling pathway involved in phagocytosis
          response to drug
          positive regulation of tyrosine phosphorylation of STAT protein
          response to amino acid
          regulation of mast cell degranulation
          negative regulation of MAP kinase activity
          positive regulation of phosphatidylinositol 3-kinase activity
          innate immune response
          regulation of erythrocyte differentiation
          protein autophosphorylation
          ephrin receptor signaling pathway
          response to axon injury
          cytokine secretion
          regulation of cytokine secretion
          regulation of inflammatory response
          negative regulation of immune response
          B cell receptor signaling pathway
          regulation of B cell receptor signaling pathway
          positive regulation of B cell receptor signaling pathway
          leukocyte migration
          positive regulation of cellular component movement
          regulation of release of sequestered calcium ion into cytosol
          positive regulation of glial cell proliferation
          positive regulation of Fc receptor mediated stimulatory signaling pathway
          JAK-STAT cascade involved in growth hormone signaling pathway
          positive regulation of stress-activated protein kinase signaling cascade
          regulation of ERK1 and ERK2 cascade
          negative regulation of ERK1 and ERK2 cascade
          positive regulation of oligodendrocyte progenitor proliferation
          negative regulation of mast cell proliferation
          positive regulation of mast cell proliferation
          cellular response to retinoic acid
          regulation of monocyte chemotaxis
          regulation of platelet aggregation
          dendritic cell differentiation
          negative regulation of intracellular signal transduction
          positive regulation of dendritic cell apoptotic process
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          receptor signaling protein tyrosine kinase activity
          receptor binding
          platelet-derived growth factor receptor binding
          integrin binding
          protein binding
          ATP binding
          SH3 domain binding
          ubiquitin protein ligase binding
          gamma-tubulin binding
          glycosphingolipid binding
          ion channel binding
          phosphoprotein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0