Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PDE4B SIRNA Assays ›
          Gene Symbol:
          PDE4B
          Gene Name:
          phosphodiesterase 4B
          Gene Aliases:
          DPDE4, PDEIVB
          Chromosome Location:
          Chr.1: 65792510 - 66374579 on Build GRCh38
          UniGene ID:
          Hs.198072
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 143737

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PDE4B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001037339.2 NP_001032416.1 3 691
          NM_001037340.2 NP_001032417.1 8 1171
          NM_001037341.1 NP_001032418.1 10 1116
          NM_001297440.1 NP_001284369.1 9 877
          NM_001297441.1 NP_001284370.1 9 770
          NM_001297442.1 NP_001284371.1 3 695
          NM_002600.3 NP_002591.2 10 1071
          XM_005270924.3 XP_005270981.1 4 362
          XM_005270925.2 XP_005270982.1 4 403
          XM_006710680.3 XP_006710743.1 4 428
          XM_011541566.1 XP_011539868.1 8 934
          XM_017001445.1 XP_016856934.1 7 523
          XM_017001446.1 XP_016856935.1 7 1121
          GenBank mRNA AB209081.1 - 8 1080
          AK289969.1 - 10 1071
          AK290006.1 - 8 1052
          AK290206.1 - 3 686
          AK294441.1 - 3 386
          BC101480.1 - 10 1011 8068986
          BC105040.1 - 10 1011 8144043
          CR749667.1 - 9 876
          EF595686.1 - 3 226
          KJ534916.1 - 9 925
          KJ534917.1 - 8 700
          KJ535082.1 - 8 686
          L12686.1 - 3 603
          L20966.1 - 10 1063
          M97515.1 - 3 690
          U85048.1 - 8 880

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_017031, U95748, M28413, AF202732, M25350, and others.
          This siRNA also targets the following transcripts in Mouse: NM_019840, NM_001177980, NM_001177981, NM_001177982, AF326555, and others.

          Panther Classification:

          Molecular Function -

          phosphodiesterase

          Gene Ontology Categories:

          Function(s) Process(es)

          neutrophil homeostasis
          cAMP catabolic process
          neutrophil chemotaxis
          positive regulation of interferon-gamma production
          positive regulation of interleukin-2 production
          cellular response to drug
          T cell receptor signaling pathway
          leukocyte migration
          cellular response to lipopolysaccharide
          cellular response to epinephrine stimulus
          regulation of cardiac muscle cell contraction
          regulation of high voltage-gated calcium channel activity
          negative regulation of relaxation of cardiac muscle
          3',5'-cyclic-AMP phosphodiesterase activity
          cAMP binding
          gamma-tubulin binding
          ion channel binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-mc8hj:80/100.66.75.242:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0