Search
Search

PDE7A| Gene Symbol: PDE7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242318.2 | NP_001229247.1 | 4 | 579 | ||
| NM_002603.3 | NP_002594.1 | 4 | 428 | |||
| XM_011517540.2 | XP_011515842.1 | 5 | 373 | |||
| XM_017013538.1 | XP_016869027.1 | 4 | 940 | |||
| GenBank mRNA | AF332652.1 | - | 4 | 353 | ||
| AK290801.1 | - | 4 | 387 | |||
| AK292680.1 | - | 4 | 376 | |||
| AY266364.1 | - | 4 | 254 | |||
| BC126360.1 | - | 4 | 420 | 8992076 | ||
| DB146108.1 | - | 4 | 376 | |||
| U67932.1 | - | 4 | 431 | |||