Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261816.1 | NP_001248745.1 | 5 | 696 | ||
NM_002613.4 | NP_002604.1 | 5 | 696 | |||
XM_011522521.2 | XP_011520823.1 | 5 | 4384 | |||
XM_017023271.1 | XP_016878760.1 | 5 | 4389 | |||
XM_017023272.1 | XP_016878761.1 | 5 | 945 | |||
XM_017023273.1 | XP_016878762.1 | 5 | 654 | |||
XM_017023274.1 | XP_016878763.1 | 6 | 792 | |||
GenBank mRNA | AB209835.1 | - | 5 | 663 | ||
AF017995.1 | - | 5 | 627 | |||
AJ785968.1 | - | 4 | 1330 | |||
AK131060.1 | - | 5 | 630 | |||
AK222581.1 | - | 5 | 639 | |||
BC012103.1 | - | 5 | 656 | 4561371 | ||
BI830733.1 | - | 5 | 698 | |||
CR536517.1 | - | 5 | 547 | |||
Y15056.1 | - | 5 | 626 |