Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTL6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004301.4 | NP_004292.1 | 3 | 470 | ||
NM_177989.3 | NP_817126.1 | 3 | 326 | |||
NM_178042.3 | NP_829888.1 | 3 | 518 | |||
GenBank mRNA | AA256726.1 | - | 3 | 441 | ||
AB015907.1 | - | 3 | 363 | |||
AB060168.1 | - | 3 | 295 | |||
AB061315.1 | - | 3 | 440 | |||
AF041474.1 | - | 3 | 393 | |||
AK021756.1 | - | 3 | 386 | |||
AK098691.1 | - | 3 | 424 | |||
AK223212.1 | - | 3 | 391 | |||
AL136608.1 | - | 3 | 410 | |||
BC000949.2 | - | 3 | 268 | 3445923 | ||
BC001391.2 | - | 3 | 311 | 3050818 | ||
BC036371.1 | - | 3 | 409 | 4827785 | ||
CR533529.1 | - | 3 | 257 | |||
DB041143.1 | - | 3 | 470 |