Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEPT2 SIRNA Assays ›
Gene Symbol:
SEPT2
Gene Name:
septin 2
Gene Aliases:
DIFF6, NEDD-5, NEDD5, Pnutl3, hNedd5
Chromosome Location:
Chr.2: 241315187 - 241354027 on Build GRCh38
UniGene ID:
Hs.721234
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
143878
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEPT2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001008491.2
NP_001008491.1
14
1825
NM_001008492.2
NP_001008492.1
14
1507
NM_001282972.1
NP_001269901.1
14
1556
NM_001282973.1
NP_001269902.1
17
1780
NM_001321029.1
NP_001307958.1
14
1541
NM_001321030.1
NP_001307959.1
14
1539
NM_001321031.1
NP_001307960.1
15
1642
NM_001321032.1
NP_001307961.1
13
1446
NM_001321033.1
NP_001307962.1
15
1658
NM_001321034.1
NP_001307963.1
15
1657
NM_001321035.1
NP_001307964.1
14
1523
NM_004404.4
NP_004395.1
13
1422
NM_006155.2
NP_006146.1
13
1724
XM_011511237.1
XP_011509539.1
13
1388
XM_011511238.1
XP_011509540.1
12
1256
XM_011511240.1
XP_011509542.1
12
1265
XM_017004191.1
XP_016859680.1
15
1617
XM_017004192.1
XP_016859681.1
15
1641
XM_017004205.1
XP_016859694.1
15
1596
XM_017004206.1
XP_016859695.1
14
1497
GenBank mRNA
AF038404.1
-
13
1361
AK294563.1
-
14
1464
BC014455.1
-
14
1704
4906575
BX648000.1
-
14
1441
CN282754.1
-
2
227
D28540.1
-
12
1343
D63878.1
-
13
1541
Back To Top
More Information
Panther Classification:
Molecular Function -
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of L-glutamate transport
mitotic nuclear division
smoothened signaling pathway
neuron projection development
regulation of protein localization
cilium assembly
regulation of catalytic activity
cell division
cell-cell adhesion
protein binding
GTP binding
enzyme regulator activity
protein complex scaffold
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0