Search
Search

NARS| Gene Symbol: NARS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004539.3 | NP_004530.1 | 8 | 978 | ||
| XM_005266700.1 | XP_005266757.1 | 8 | 756 | |||
| GenBank mRNA | AJ000334.1 | - | 8 | 823 | ||
| AK129897.1 | - | 8 | 781 | |||
| AK222835.1 | - | 8 | 825 | |||
| AK293985.1 | - | 7 | 758 | |||
| AK294364.1 | - | 7 | 754 | |||
| AK294763.1 | - | 8 | 809 | |||
| BC001687.1 | - | 8 | 827 | 2987840 | ||
| D84273.1 | - | 8 | 782 | |||