Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MEF2A SIRNA Assays ›
Gene Symbol:
MEF2A
Gene Name:
myocyte enhancer factor 2A
Gene Aliases:
ADCAD1, RSRFC4, RSRFC9, mef2
Chromosome Location:
Chr.15: 99565455 - 99716488 on Build GRCh38
UniGene ID:
Hs.268675
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
144067
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MEF2A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_005587.3
NP_005578.2
5
709
XM_005254914.2
XP_005254971.1
5
674
XM_005254916.3
XP_005254973.1
5
674
XM_011521571.2
XP_011519873.1
5
674
XM_011521572.2
XP_011519874.1
5
619
XM_011521573.2
XP_011519875.1
5
649
XM_011521576.2
XP_011519878.1
4
563
XM_011521577.2
XP_011519879.1
4
497
XM_011521578.2
XP_011519880.1
4
1156
XM_011521579.2
XP_011519881.1
4
573
XM_011521581.2
XP_011519883.1
5
937
XM_011521582.2
XP_011519884.1
6
1142
XM_011521585.2
XP_011519887.1
4
733
XM_011521590.2
XP_011519892.1
5
674
XM_017022190.1
XP_016877679.1
4
563
XM_017022191.1
XP_016877680.1
4
555
XM_017022192.1
XP_016877681.1
4
552
XM_017022193.1
XP_016877682.1
5
747
XM_017022194.1
XP_016877683.1
4
563
XM_017022195.1
XP_016877684.1
4
420
XM_017022196.1
XP_016877685.1
6
1063
XM_017022197.1
XP_016877686.1
5
2854
XM_017022198.1
XP_016877687.1
6
1063
XM_017022199.1
XP_016877688.1
4
1352
XM_017022200.1
XP_016877689.1
5
1134
XM_017022201.1
XP_016877690.1
4
757
XM_017022203.1
XP_016877692.1
5
937
GenBank mRNA
BC013437.2
-
5
627
3896491
KU178181.1
-
3
308
KU178182.1
-
3
308
X63381.1
-
4
536
X68505.1
-
5
722
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001014035, XM_008759542, XM_008759540, DQ323505, GU646868, and others.
This siRNA also targets the following transcripts in Mouse: NM_001291191, NM_001291192, AK040227, BC096598, AK153751.
Panther Classification:
Molecular Function -
MADS box transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
mitochondrial genome maintenance
negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
transcription, DNA-templated
transcription from RNA polymerase II promoter
apoptotic process
heart development
muscle organ development
positive regulation of cardiac muscle hypertrophy
positive regulation of transcription from RNA polymerase II promoter
mitochondrion distribution
dendrite morphogenesis
positive regulation of muscle cell differentiation
ventricular cardiac myofibril assembly
cardiac conduction
ERK5 cascade
cellular response to calcium ion
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
RNA polymerase II transcription coactivator activity
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
protein kinase binding
activating transcription factor binding
histone acetyltransferase binding
histone deacetylase binding
sequence-specific DNA binding
SMAD binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0