Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CD200 SIRNA Assays ›
Gene Symbol:
CD200
Gene Name:
CD200 molecule
Gene Aliases:
MOX1, MOX2, MRC, OX-2
Chromosome Location:
Chr.3: 112332224 - 112362812 on Build GRCh38
UniGene ID:
Hs.79015
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
144150
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CD200
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001004196.3
NP_001004196.2
4
643
NM_001318826.1
NP_001305755.1
3
568
NM_001318828.1
NP_001305757.1
2
486
NM_001318830.1
NP_001305759.1
2
486
NM_005944.6
NP_005935.4
3
568
XM_005247481.4
XP_005247538.1
4
817
XM_005247482.2
XP_005247539.1
3
447
XM_005247485.2
XP_005247542.1
3
758
XM_005247486.2
XP_005247543.1
2
255
XM_005247488.2
XP_005247545.1
2
365
XM_017006452.1
XP_016861941.1
4
1144
XM_017006453.1
XP_016861942.1
2
410
XM_017006454.1
XP_016861943.1
4
1133
XM_017006455.1
XP_016861944.1
3
768
XM_017006456.1
XP_016861945.1
4
494
XM_017006457.1
XP_016861946.1
3
447
XM_017006458.1
XP_016861947.1
4
1139
XM_017006459.1
XP_016861948.1
3
758
GenBank mRNA
AF063591.1
-
3
359
AK074972.1
-
3
333
AK129797.1
-
2
255
AK293399.1
-
2
255
AK297194.1
-
4
434
AK312563.1
-
3
449
AL832529.1
-
3
353
AY603771.1
-
2
302
BC022522.1
-
3
361
4794050
BC031103.1
-
3
337
5301702
BI547787.1
-
4
439
DA544134.1
-
3
446
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
cell recognition
negative regulation of macrophage activation
regulation of immune response
receptor activity
receptor binding
protein binding
protein homodimerization activity
cell adhesion molecule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0