Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MBD1 SIRNA Assays ›
          Gene Symbol:
          MBD1
          Gene Name:
          methyl-CpG binding domain protein 1
          Gene Aliases:
          CXXC3, PCM1, RFT
          Chromosome Location:
          Chr.18: 50266882 - 50281774 on Build GRCh38
          UniGene ID:
          Hs.405610
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 144322

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MBD1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001204136.1 NP_001191065.1 16 2357
          NM_001204142.1 NP_001191071.1 14 2424
          NM_001204143.1 NP_001191072.1 14 1825
          NM_001204151.2 NP_001191080.1 15 2319
          NM_001323942.1 NP_001310871.1 17 2432
          NM_001323947.1 NP_001310876.1 17 2387
          NM_001323949.1 NP_001310878.1 18 2484
          NM_001323950.1 NP_001310879.1 17 2381
          NM_001323952.1 NP_001310881.1 16 2186
          NM_001323954.1 NP_001310883.1 14 2120
          NM_002384.2 NP_002375.1 15 2123
          NM_015844.2 NP_056669.2 16 2261
          NM_015845.3 NP_056670.2 14 2150
          NM_015846.3 NP_056671.2 17 2429
          NM_015847.3 NP_056723.2 16 2282
          XM_005258271.2 XP_005258328.1 16 3826
          XM_006722456.2 XP_006722519.1 17 2172
          XM_011525991.1 XP_011524293.1 16 2041
          XM_011525993.2 XP_011524295.1 18 2251
          XM_011525994.2 XP_011524296.1 18 2244
          XM_011525999.1 XP_011524301.1 15 1966
          XM_011526001.1 XP_011524303.1 15 1936
          XM_011526002.1 XP_011524304.1 16 3763
          XM_011526003.2 XP_011524305.1 17 2113
          XM_011526006.1 XP_011524308.1 14 1791
          XM_011526007.1 XP_011524309.1 15 3688
          XM_017025751.1 XP_016881240.1 17 2127
          XM_017025752.1 XP_016881241.1 16 2059
          XM_017025753.1 XP_016881242.1 16 2097
          XM_017025756.1 XP_016881245.1 16 2034
          XM_017025757.1 XP_016881246.1 17 3856
          XM_017025758.1 XP_016881247.1 18 2206
          XM_017025759.1 XP_016881248.1 18 2199
          XM_017025760.1 XP_016881249.1 17 2169
          XM_017025762.1 XP_016881251.1 17 2124
          XM_017025763.1 XP_016881252.1 16 3778
          XM_017025764.1 XP_016881253.1 17 2106
          XM_017025765.1 XP_016881254.1 15 1929
          XM_017025766.1 XP_016881255.1 17 2083
          XM_017025767.1 XP_016881256.1 17 2076
          XM_017025769.1 XP_016881258.1 16 2038
          XM_017025770.1 XP_016881259.1 15 3658
          XM_017025771.1 XP_016881260.1 16 2001
          XM_017025773.1 XP_016881262.1 14 3589
          XM_017025774.1 XP_016881263.1 15 1939
          XM_017025776.1 XP_016881265.1 14 3520
          GenBank mRNA AF078831.1 - 13 1713
          AJ564845.1 - 14 2126
          AK225889.1 - 16 2267
          AK300759.1 - 14 1825
          BC033242.1 - 16 1987 5432943
          Y10746.1 - 16 1984

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          methylation-dependent chromatin silencing
          transcription from RNA polymerase II promoter
          heart development
          aging
          response to fluoxetine
          neuron differentiation
          response to nutrient levels
          response to estradiol
          response to cocaine
          regulation of DNA methylation
          negative regulation of transcription, DNA-templated
          negative regulation of astrocyte differentiation
          DNA binding
          chromatin binding
          protein binding
          zinc ion binding
          methyl-CpG binding
          double-stranded methylated DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lq2bv:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0