siRNA Details
Gene Symbol: NOTCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017617.4 | NP_060087.3 | 7 | 1127 | ||
XM_011518717.2 | XP_011517019.2 | 4 | 419 | |||
GenBank mRNA | AF308602.1 | - | 7 | 1127 | ||
CN431067.1 | - | 3 | 370 | |||
M73980.1 | - | 7 | 1127 |

More Information
Panther Classification:
Molecular Function -




