Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLEC SIRNA Assays ›
Gene Symbol:
PLEC
Gene Name:
plectin
Gene Aliases:
EBS1, EBSMD, EBSND, EBSO, EBSOG, EBSPA, HD1, LGMD2Q, PCN, PLEC1, PLEC1b, PLTN
Chromosome Location:
Chr.8: 143915147 - 143976800 on Build GRCh38
UniGene ID:
Hs.434248
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
144453
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PLEC
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000445.4
NP_000436.2
33
14736
NM_201378.3
NP_958780.1
32
14621
NM_201379.2
NP_958781.1
32
14689
NM_201380.3
NP_958782.1
32
15187
NM_201381.2
NP_958783.1
32
14584
NM_201382.3
NP_958784.1
32
14674
NM_201383.2
NP_958785.1
32
14627
NM_201384.2
NP_958786.1
32
14735
XM_005250976.3
XP_005251033.1
33
15229
XM_005250978.3
XP_005251035.1
33
14754
XM_005250979.4
XP_005251036.1
33
14713
XM_005250980.4
XP_005251037.1
33
14798
XM_005250981.3
XP_005251038.1
33
14635
XM_005250982.3
XP_005251039.1
33
14628
XM_005250983.2
XP_005251040.1
32
14552
XM_005250984.4
XP_005251041.1
33
14617
XM_006716588.3
XP_006716651.1
34
14806
XM_006716589.2
XP_006716652.1
33
14567
XM_006716590.3
XP_006716653.1
33
14677
XM_011517130.2
XP_011515432.1
33
14742
XM_011517131.2
XP_011515433.1
32
14661
XM_011517132.2
XP_011515434.1
32
11369
GenBank mRNA
AY480044.1
-
32
14693
AY480045.1
-
32
14564
AY480046.1
-
32
14689
AY480047.1
-
32
15187
AY480048.1
-
32
14584
AY480049.1
-
32
14613
AY480050.1
-
32
14627
AY480051.1
-
32
14735
BC007597.1
-
1
4052
BC013206.1
-
1
2001
U53204.1
-
33
14738
Back To Top
More Information
Panther Classification:
Molecular Function -
intermediate filament binding protein
Gene Ontology Categories:
Function(s)
Process(es)
hemidesmosome assembly
cell-cell adhesion
actin binding
protein binding
structural constituent of muscle
ankyrin binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0