Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LTBP1 SIRNA Assays ›
Gene Symbol:
LTBP1
Gene Name:
latent transforming growth factor beta binding protein 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 32947129 - 33399509 on Build GRCh38
UniGene ID:
Hs.619315
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
144461
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LTBP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000627.3
NP_000618.3
1, 2
358
NM_001166264.1
NP_001159736.1
1, 2
358
NM_001166265.1
NP_001159737.1
1, 2
358
NM_001166266.1
NP_001159738.1
1, 2
358
NM_206943.2
NP_996826.2
5, 6
1218
XM_005264317.3
XP_005264374.1
5, 6
1220
XM_005264318.3
XP_005264375.1
5, 6
1220
XM_011532853.2
XP_011531155.1
5, 6
1391
XM_011532855.2
XP_011531157.1
5, 6
1220
XM_011532856.2
XP_011531158.1
5, 6
1220
XM_011532857.2
XP_011531159.1
5, 6
1220
XM_011532858.2
XP_011531160.1
5, 6
1220
XM_011532859.2
XP_011531161.1
5, 6
1220
XM_011532861.2
XP_011531163.1
5, 6
1220
XM_011532862.2
XP_011531164.1
5, 6
1220
XM_017004108.1
XP_016859597.1
5, 6
1220
XM_017004109.1
XP_016859598.1
5, 6
1220
XM_017004110.1
XP_016859599.1
5, 6
1220
GenBank mRNA
AB208801.1
-
1, 2
316
AF489528.1
-
1, 2
217
BC130289.1
-
1, 2
298
40146320
BC144127.1
-
1, 2
298
BC144128.1
-
1, 2
298
9052648
BM147075.1
-
1, 2
360
BX472809.1
-
1, 2
346
BX492839.1
-
1, 2
438
CD657511.1
-
3, 4
569
M34057.1
-
1, 2
307
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_021587, M55431.
Panther Classification:
Molecular Function -
extracellular matrix structural protein
Gene Ontology Categories:
Function(s)
Process(es)
ventricular septum development
transmembrane receptor protein serine/threonine kinase signaling pathway
sequestering of TGFbeta in extracellular matrix
aorta development
coronary vasculature development
transforming growth factor beta-activated receptor activity
calcium ion binding
protein binding
transforming growth factor beta binding
microfibril binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-c4z4p:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0