Search
Search

HK2| Gene Symbol: HK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000189.4 | NP_000180.2 | 18 | 4858 | ||
| XM_005264280.1 | XP_005264337.1 | 18 | 3401 | |||
| XM_011532807.1 | XP_011531109.1 | 17 | 3305 | |||
| XM_017003945.1 | XP_016859434.1 | 17 | 3242 | |||
| GenBank mRNA | AF148513.1 | - | 18 | 3381 | ||
| AK290357.1 | - | 18 | 3361 | |||
| BC064369.1 | - | 18 | 3374 | 5586773 | ||
| CR749202.1 | - | 18 | 3038 | |||